ENA447957843 Enamine

3-methylthiophene-2-sulfonamide

  • MDL number MFCD18817151
  • Cas Number : 81417-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208181

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208181
Availability details for
Quantity
  • Availability
  • Add to Cart