ENA447957845 Enamine

2-(4-chlorophenyl)-2,2-difluoroethan-1-amine

  • MDL number MFCD20699836
  • Cas Number : 730913-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205918

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205918
Availability details for
Quantity
  • Availability
  • Add to Cart