ENA447957853 Enamine

1-(5-bromothiophen-2-yl)cyclobutane-1-carbonitrile

  • MDL number MFCD26653601
  • Cas Number : 1514316-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205943

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205943
Availability details for
Quantity
  • Availability
  • Add to Cart