ENA447957854 Enamine

ethyl 5-(4-chlorophenyl)-1-methyl-1H-1,2,4-triazole-3-carboxylate

  • MDL number MFCD19482214
  • Cas Number : 1283530-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208201

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208201
Availability details for
Quantity
  • Availability
  • Add to Cart