ENA447957860 Enamine

1-[1-(5-bromothiophen-2-yl)cyclobutyl]methanamine

  • MDL number MFCD28397538
  • Cas Number : 1803604-58-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208223
Availability details for
Quantity
  • Availability
  • Add to Cart