ENA447957861 Enamine

3H,4H,7H-imidazo[4,5-d][1,2,3]triazin-4-one

  • MDL number MFCD19216826
  • Cas Number : 4656-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-185
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208225

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P312,P330,P363,P501
MSDS https://www.enaminestore.com/catalog/EN300-208225
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart