ENA447957863 Enamine

3-(3-ethyl-1,2-oxazol-5-yl)propan-1-ol

  • MDL number MFCD26820544
  • Cas Number : 1566151-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205948

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205948
Availability details for
Quantity
  • Availability
  • Add to Cart