ENA447958114 Enamine

1-(2,4,5-trichlorophenyl)methanamine hydrochloride

  • MDL number MFCD28397541
  • Cas Number : 1803571-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208246

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208246
Availability details for
Quantity
  • Availability
  • Add to Cart