ENA447958116 Enamine

ethyl 3-(chlorosulfonyl)-4,4,4-trifluorobutanoate

  • MDL number MFCD28397543
  • Cas Number : 1803598-32-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208280

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208280
Availability details for
Quantity
  • Availability
  • Add to Cart