ENA447958120 Enamine

1-(2-chlorophenyl)-1-(1H-pyrazol-1-yl)propan-2-amine dihydrochloride

  • MDL number MFCD28397548
  • Cas Number : 1803589-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208313

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208313
Availability details for
Quantity
  • Availability
  • Add to Cart