ENA447958126 Enamine

1-(but-3-en-1-yl)piperidin-4-amine dihydrochloride

  • MDL number MFCD28397555
  • Cas Number : 1803612-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208351

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208351
Availability details for
Quantity
  • Availability
  • Add to Cart