ENA447958129 Enamine

2-(4-methylpiperidin-4-yl)-1,3-benzothiazole hydrochloride

  • MDL number MFCD28397560
  • Cas Number : 1803604-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208394

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208394
Availability details for
Quantity
  • Availability
  • Add to Cart