ENA447958130 Enamine

methyl 2-(methylamino)-3-nitropyridine-4-carboxylate

  • MDL number MFCD28397561
  • Cas Number : 1803593-34-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208410

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208410
Availability details for
Quantity
  • Availability
  • Add to Cart