ENA447958131 Enamine

3-(oxolan-2-yl)morpholine

  • MDL number MFCD28397562
  • Cas Number : 1803561-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208425

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208425
Availability details for
Quantity
  • Availability
  • Add to Cart