ENA447958137 Enamine

1-(1H-imidazol-2-yl)-4,4-dimethylpentan-1-amine dihydrochloride

  • MDL number MFCD28397568
  • Cas Number : 1803605-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208468

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208468
Availability details for
Quantity
  • Availability
  • Add to Cart