ENA447958152 Enamine

2-methoxy-4-azatricyclo[4.2.1.0,3,7]nonane hydrochloride

  • MDL number MFCD28397586
  • Cas Number : 1803605-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208657

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208657
Availability details for
Quantity
  • Availability
  • Add to Cart