ENA447958159 Enamine

methyl({[4-(2,2,2-trifluoroethoxy)phenyl]methyl})amine hydrochloride

  • MDL number MFCD28397593
  • Cas Number : 1803600-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208756

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208756
Availability details for
Quantity
  • Availability
  • Add to Cart