ENA447958160 Enamine

4,4,4-trifluoro-3-(1-methyl-1H-imidazol-2-yl)butanoic acid hydrochloride

  • MDL number MFCD28397594
  • Cas Number : 1803570-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208798

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208798
Availability details for
Quantity
  • Availability
  • Add to Cart