ENA447958161 Enamine

7-chloro-3,4-dihydro-2H-1-benzopyran-4-amine hydrochloride

  • MDL number MFCD28397595
  • Cas Number : 191608-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208806

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208806
Availability details for
Quantity
  • Availability
  • Add to Cart