ENA447958164 Enamine

{3-[(acetyloxy)imino]-2,3-dihydro-1H-isoindol-1-ylidene}amino acetate

  • Cas Number : 1089332-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208853

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208853
Availability details for
Quantity
  • Availability
  • Add to Cart