ENA447958171 Enamine

2-(3,5-difluorophenoxy)-2-methylpropan-1-amine hydrochloride

  • MDL number MFCD28397605
  • Cas Number : 1803601-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208884

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208884
Availability details for
Quantity
  • Availability
  • Add to Cart