ENA447958172 Enamine

2-amino-2-methyl-3-(methylsulfanyl)propanenitrile hydrochloride

  • MDL number MFCD28397606
  • Cas Number : 1803593-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208886

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208886
Availability details for
Quantity
  • Availability
  • Add to Cart