ENA447958175 Enamine

methyl 4-(aminomethyl)piperidine-1-carboxylate hydrochloride

  • MDL number MFCD28397611
  • Cas Number : 1803572-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208953

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208953
Availability details for
Quantity
  • Availability
  • Add to Cart