ENA447958176 Enamine

5-(furan-2-yl)-1,2,3,6-tetrahydropyridine hydrochloride

  • MDL number MFCD28397612
  • Cas Number : 1803570-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208955

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208955
Availability details for
Quantity
  • Availability
  • Add to Cart