ENA447958178 Enamine

2-(4-bromophenoxy)-3-methylbutanenitrile

  • MDL number MFCD28397614
  • Cas Number : 1803598-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208964

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208964
Availability details for
Quantity
  • Availability
  • Add to Cart