ENA447958179 Enamine

1-(3-chloro-5-methanesulfonylphenyl)methanamine hydrochloride

  • MDL number MFCD28397615
  • Cas Number : 1192347-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 239-241
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208991

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208991
Availability details for
Quantity
  • Availability
  • Add to Cart