ENA447958181 Enamine

1-methyl-3-(methylamino)pyrrolidin-2-one hydrochloride

  • MDL number MFCD28397617
  • Cas Number : 1803561-70-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209048

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209048
Availability details for
Quantity
  • Availability
  • Add to Cart