ENA447958183 Enamine

(2R)-1-(methylamino)propan-2-ol hydrochloride

  • MDL number MFCD28397619
  • Cas Number : 929533-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209068

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209068
Availability details for
Quantity
  • Availability
  • Add to Cart