ENA447958184 Enamine

3-(1H-imidazol-2-yl)-3-phenylpropanoic acid hydrochloride

  • MDL number MFCD28397620
  • Cas Number : 1803570-27-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209073

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209073
Availability details for
Quantity
  • Availability
  • Add to Cart