ENA447958185 Enamine

5-(aminomethyl)-4H,5H,6H,7H-pyrazolo[1,5-a]pyridine-3-carboxamide dihydrochloride

  • MDL number MFCD28397622
  • Cas Number : 1803593-49-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209101

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209101
Availability details for
Quantity
  • Availability
  • Add to Cart