ENA447958187 Enamine

{[(2S,4S)-4-fluoro-1-(5-methyl-1,3,4-oxadiazol-2-yl)pyrrolidin-2-yl]methyl}(methyl)amine dihydrochloride

  • MDL number MFCD31566981
  • Cas Number : 1807939-09-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209111

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209111
Availability details for
Quantity
  • Availability
  • Add to Cart