ENA447958189 Enamine

2-amino-2-(4-bromophenyl)propan-1-ol hydrochloride

  • MDL number MFCD28397626
  • Cas Number : 1803598-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209115

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209115
Availability details for
Quantity
  • Availability
  • Add to Cart