ENA447958195 Enamine

1,2,5,6-tetrahydro-3,4'-bipyridine hydrochloride

  • MDL number MFCD28397630
  • Cas Number : 1803600-61-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209129

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209129
Availability details for
Quantity
  • Availability
  • Add to Cart