ENA447958224 Enamine

(piperidin-2-yl)methyl N-(propan-2-yl)carbamate hydrochloride

  • MDL number MFCD28397639
  • Cas Number : 1803604-09-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209204

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209204
Availability details for
Quantity
  • Availability
  • Add to Cart