ENA447958225 Enamine

4-amino-2-chloropyridine-3-carboxamide hydrochloride

  • MDL number MFCD28397640
  • Cas Number : 1803589-19-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209206

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209206
Availability details for
Quantity
  • Availability
  • Add to Cart