ENA447958233 Enamine

2-{[(tert-butoxy)carbonyl]amino}-3-(piperidin-1-yl)propanoic acid hydrochloride

  • MDL number MFCD28397648
  • Cas Number : 1803562-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209258

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209258
Availability details for
Quantity
  • Availability
  • Add to Cart