ENA447958239 Enamine

[2-(azepan-1-yl)ethyl](cyclopropylmethyl)amine hydrochloride

  • MDL number MFCD28397650
  • Cas Number : 1803572-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209295

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209295
Availability details for
Quantity
  • Availability
  • Add to Cart