ENA447958267 Enamine

4-(5,5-dimethyl-4,5-dihydro-1,3-thiazol-2-yl)piperazine-1-carbaldehyde

  • MDL number MFCD28397666
  • Cas Number : 1803586-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209473

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209473
Availability details for
Quantity
  • Availability
  • Add to Cart