ENA447958269 Enamine

6-(4-aminopiperidin-1-yl)pyridine-3-carboxylic acid dihydrochloride

  • MDL number MFCD28397668
  • Cas Number : 1803594-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209485

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209485
Availability details for
Quantity
  • Availability
  • Add to Cart