ENA447958272 Enamine

4-(1,3,4-oxadiazol-2-yl)piperidine; trifluoroacetic acid

  • MDL number MFCD28397672
  • Cas Number : 1803604-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data CF3COOH
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209529

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209529
Availability details for
Quantity
  • Availability
  • Add to Cart