ENA447958274 Enamine

2-(4-chlorophenyl)-5-methoxypentanenitrile

  • MDL number MFCD28397674
  • Cas Number : 1803594-33-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209538
Availability details for
Quantity
  • Availability
  • Add to Cart