ENA447958275 Enamine

1-(2-isocyanatoethyl)-4-(trifluoromethoxy)benzene

  • MDL number MFCD28397675
  • Cas Number : 730981-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209548

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209548
Availability details for
Quantity
  • Availability
  • Add to Cart