ENA447958278 Enamine

1-[(2S,4S)-4-fluoro-1-[(5-methyl-1,2-oxazol-4-yl)methyl]pyrrolidin-2-yl]methanamine dihydrochloride

  • MDL number MFCD31566984
  • Cas Number : 1820583-09-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209618

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209618
Availability details for
Quantity
  • Availability
  • Add to Cart