ENA447958280 Enamine

[2-(methylamino)ethyl][2-(piperidin-1-yl)ethyl]amine trihydrochloride

  • MDL number MFCD28397680
  • Cas Number : 1803588-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 3HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209623
Availability details for
Quantity
  • Availability
  • Add to Cart