ENA447958282 Enamine

(piperidin-2-yl)methyl morpholine-4-carboxylate hydrochloride

  • MDL number MFCD28397683
  • Cas Number : 1803598-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209631

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209631
Availability details for
Quantity
  • Availability
  • Add to Cart