ENA447958285 Enamine

N-(2-hydroxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1-sulfonamide

  • MDL number MFCD28397687
  • Cas Number : 1014613-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209647

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209647
Availability details for
Quantity
  • Availability
  • Add to Cart