ENA447958287 Enamine

4-{[2-(methylamino)ethoxy]methyl}benzonitrile hydrochloride

  • MDL number MFCD28397691
  • Cas Number : 1803606-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210066

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210066
Availability details for
Quantity
  • Availability
  • Add to Cart