ENA447958291 Enamine

1-{3-oxabicyclo[3.1.0]hexan-6-yl}methanamine hydrochloride

  • MDL number MFCD28397695
  • Cas Number : 1803598-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210089

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210089
Availability details for
Quantity
  • Availability
  • Add to Cart