ENA447958294 Enamine

1-[2-(4-aminopiperidin-1-yl)acetyl]pyrrolidin-2-one dihydrochloride

  • MDL number MFCD28397698
  • Cas Number : 1803586-67-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210094

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210094
Availability details for
Quantity
  • Availability
  • Add to Cart