ENA447958295 Enamine

1-(2-aminoethyl)-1H-1,2,4-triazole-3-carbonitrile hydrochloride

  • MDL number MFCD28397699
  • Cas Number : 1803598-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210104

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210104
Availability details for
Quantity
  • Availability
  • Add to Cart